Human recombinant omentin (from CHO cells)
Fournisseur: ProSci Inc.
90-249
PRSI90-249EA 400 EUR InStock
PRSI90-249
Human recombinant omentin (from CHO cells)
Protéines et Peptides
Omentin (Intelectin-1) is highly expressed in omental adipose tissue, small intestine and various other tissues. Omentin has no effect on basal glucose uptake but enhances insulin-stimulated glucose uptake in adipocytes. It increases AKT phosphorylation in the absence and presence of insulin. It was suggested to play a role in the defence system against microorganisms by recognising carbohydrate chains of pathogens and bacterial components containing galactofuranosyl residues.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité