Human Recombinant CD137 (from HEK293 cells)
Fournisseur: ProSci Inc.
90-252
PRSI90-252EA 567 EUR InStock
PRSI90-252
Human Recombinant CD137 (from HEK293 cells)
Protéines et Peptides
CD137 (4-1BB) is a member of the tumor necrosis factor (TNF) receptor family. CD137 is expressed by activated T cells, dendritic cells, NK cells, granulocytes and cells of blood vessel walls at sites of inflammation. CD137 has costimulatory activity for activated T cells. Crosslinking of CD137 enhances T cell proliferation, IL-2 secretion survival and cytolytic activity. CD137 was shown to enhance immune activity to eliminate tumours in mice.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité