Human recombinant Angiocidin (from E. coli)
Fournisseur: ProSci Inc.
90-253
PRSI90-253EA 654 EUR InStock
PRSI90-253
Human recombinant Angiocidin (from E. coli)
Protéines et Peptides
Angiocidin shows sequence similarity with proteasome components and is also being referred to as 26A proteasome regulatory subunit S5A. Angiocidin shows both matrix and cytoplasmic distribution. It binds to thrombospondin-1 and alpha2beta1 integrin, and it inhibits tumour progression and angiogenesis. Angiocidin is expressed in various types of cancer, such as hepatocellular carcinoma, prostate, lung, bladder, and melanoma cancers.
Fusion-Tag: Tag Free
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité