Human recombinant IZTRAIL (soluble) (from E. coli)
Fournisseur: ProSci Inc.
90-257
PRSI90-257EA 488 EUR InStock
PRSI90-257
Human recombinant IZTRAIL (soluble) (from E. coli)
Protéines et Peptides
izTRAIL is a newly available, highly active recombinant form of soluble human TRAIL. Due to a trimerizing N-terminal isoleucine zipper (iz) motif the intrinsic trimerization of TRAIL, required for apoptosis-inducing activity of TRAIL, is enhanced when compared to non-tagged soluble human TRAIL (shTRAIL). Therefore, izTRAIL is a potent inducer of apoptosis in many human cancer cells, but not normal human hepatocytes. In addition, the half-life of izTRAIL is about eight-fold higher than the half-life of shTRAIL. These properties render izTRAIL highly suitable for both in vitro and in vivo use, particularly for studies in which investigators plan to transfer their in vitro results into an in vivo system with human cancer cells in xenotransplant settings examining susceptibility to TRAIL-induced apoptosis.
Fusion-Tag: isoleucine zipper motif Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité