Mouse recombinant darcin (from E. coli)
Fournisseur: ProSci Inc.
90-264
PRSI90-264EA 400 EUR InStock
PRSI90-264
Mouse recombinant darcin (from E. coli)
Protéines et Peptides
Darcin (also called Mup20) belongs to the family of major urinary proteins (MUPs), a subfamily of proteins found in abundance in the urine. Like all MUPs, Darcin is a member of the lipocalin family of ligand-binding proteins, and preferentially binds the volatile pheromone (S)-2-sec-butyl-4,5-dihydrothiazole (SBT). Darcin is present in the urine of male and acts as a sex pheromone that elicits female sexual attraction to a male's urine scent. Recombinant Darcin shows similar activity compared to natural Darcin.
Fusion-Tag: Tag Free
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité