Mouse recombinant periostin (from CHO cells)
Fournisseur: ProSci Inc.
90-267
PRSI90-267EA 528 EUR InStock
PRSI90-267
Mouse recombinant periostin (from CHO cells)
Protéines et Peptides
Periostin was originally isolated as an osteoblast-specific factor that functions as a cell adhesion molecule for pre-osteoblasts and is thought to be involved in osteoblast recruitment, attachment and spreading. Additionally, periostin expression has previously been shown to be significantly increased by both transforming growth factor beta1 (TGF-beta1) and bone morphogenetic protein 2 (BMP-2). Abnormal expression of Periostin is also linked to angiogenesis and metastasis in epithelial tumours.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité