Human recombinant APRIL H98 multimeric (from HEK293 cells)
Fournisseur: ProSci Inc.
90-272
PRSI90-272EA 620 EUR InStock
PRSI90-272
Human recombinant APRIL H98 multimeric (from HEK293 cells)
Protéines et Peptides
APRIL is a cytokine that belongs to the TNF superfamily and binds to TACI and BCMA. It is implicated in the regulation of tumour cell growth, is involved in monocyte/macrophage-mediated immunological processes and functions as an important survival factor for plasmablasts and bone marrow plasma cells. MultimericAPRIL™ is a high activity construct in which two trimeric APRIL ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively stimulates proliferation B cell. A basic amino acid sequence (QKQKKQ) close to the N-terminus of APRIL is required for binding to negatively charged sulphated glycosaminoglycan side chains of proteoglycans. Proteoglycans mediate binding of APRIL to tumour cells as well as primary lymphoid cells.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité