Mouse recombinant Notch2 (from CHO cells)
Fournisseur: ProSci Inc.
90-292
PRSI90-292EA 567 EUR InStock
PRSI90-292
Mouse recombinant Notch2 (from CHO cells)
Protéines et Peptides
Notch signalling pathway regulates many different cell fate decisions in both vertebrate and invertebrate species. There are 5 canonical Notch ligands in mammals: Jagged-1, Jagged-2, DLL1, DLL3 and DLL4. These can bind to the four Notch receptors Notch 1-4. It is important for pattern formation during development such as neurogenesis, angiogenesis or myogenesis and regulates T cell development and stem cell maintenance. Notch signalling is also involved in cellular processes through-out adulthood. Signalling via Notch occurs between neighbouring cells and both the receptor and its ligands are transmembrane proteins.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité