Mouse recombinant profilin toxoplasma gondii (from E. coli)
Fournisseur: ProSci Inc.
90-302
PRSI90-302EA 488 EUR InStock
PRSI90-302
Mouse recombinant profilin toxoplasma gondii (from E. coli)
Protéines et Peptides
TLR11, an innate sensor for profilin (Toxoplasma gondii), is an intracellular receptor that resides in the endoplasmic reticulum. The 12 membrane-spanning endoplasmic reticulum-resident protein UNC93B1 interacts directly with TLR11 and regulates the activation of dendritic cells in response to profilin (Toxoplasma gondii) and parasitic infection in vivo. TLR12, a previously uncharacterised TLR, also recognises profilin from Toxoplasma gondii. TLR12 is sufficient for recognition of profilin by plasmacytoid dendritic cells (pDCs), whereas TLR11 and TLR12 are both required in macrophages and conventional DCs. In contrast to TLR11, TLR12-deficient mice succumb rapidly to T. gondii infection.
Fusion-Tag: Tag Free
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité