Human recombinant FasL multimeric (from HEK293 cells)
Fournisseur: ProSci Inc.
90-310
PRSI90-310EA 553 EUR InStock
PRSI90-310
Human recombinant FasL multimeric (from HEK293 cells)
Protéines et Peptides
MultimericFasL™ is a high activity construct in which two trimeric FasL are artificially linked via the collagen domain of ACRP30. This construct very effectively mimics the natural membrane-assisted aggregation of FasL in vivo. FasL is a cytokine that binds to TNFRSF6/Fas, a receptor that transduces the apoptotic signal into cells. It is involved in cytotoxic T cell mediated apoptosis and in T cell development.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité