Mouse recombinant dectin-1 (from HEK293 cells)
Fournisseur: ProSci Inc.
90-318
PRSI90-318EA 601 EUR InStock
PRSI90-318
Mouse recombinant dectin-1 (from HEK293 cells)
Protéines et Peptides
Dectin-1 and dectin-2 are type II transmembrane proteins of the C-type lectin family. They contain single carbohydrate recognition domains (CRDs) in their extracellular region. Dectin-1 and dectin-2 are expressed in macrophages and dendritic cells. Dectin-1 recognises beta-glucans with its CRD and signals through its immunoreceptor tyrosine-based activation motif (ITAM)-like motif in the cytoplasmic domain, whereas dectin-2 recognises alpha-mannans and transduces its signal through association with the ITAM-containing Fc receptor gamma-chain.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité