Mouse recombinant Meteorin-like (from HEK293 cells)
Fournisseur: ProSci Inc.
90-330
PRSI90-330EA 448 EUR InStock
PRSI90-330
Mouse recombinant Meteorin-like (from HEK293 cells)
Protéines et Peptides
Meteorin-like (cometin; subfatin) is a potent new neurotrophic factor promoting neurite outgrowth from dorsal root ganglion cells. Meteorin-like is also a novel adipokine expressed by adipose tissue being downregulated upon caloric restriction. Meteorin-like is secreted by muscles during exercise and converts white adipose cells into brown fat tissue.
Fusion-Tag: DDDDK Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité