Human recombinant KillerTRAIL (soluble) (from E. coli)
Fournisseur: ProSci Inc.
90-335
PRSI90-335EA 762 EUR InStock
PRSI90-335
Human recombinant KillerTRAIL (soluble) (from E. coli)
Protéines et Peptides
TNF-related apoptosis-inducing ligand (TRAIL; Apo2L;CD253; TNFSF10) is a type II transmembrane protein of about 34kDa. Like most members of the tumor necrosis factor (TNF) superfamily of cytokines TRAIL can be cleaved at the cell surface by metalloproteases to form a soluble molecule. Active TRAIL forms trimers and specifically binds to five distinct receptors: TRAIL-R1 (DR4; Apo2;CD261; TNFRSF10A), TRAIL-R2 (DR5; KILLER; TRICK2A;TRICK2B; CD262; TNFRSF10B), TRAIL-R3 (DcR1;LIT; TRID; CD263; TNFRSF10C), TRAIL-R4 (DcR2;TRUNDD; CD264; TNFRSF10D), and osteoprotegerin (OPG; OCIF; TNFRSF11B). Trimerised TRAIL triggers apoptosis upon ligation of cell surface TRAIL-R1 and/or TRAIL-R2 by inducing the formation of the so-called multiprotein death-inducing signalling complex (DISC).
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité