Human recombinant PARP1 (from Insect cells (Sf21))
Fournisseur: ProSci Inc.
90-340
PRSI90-340EA 567 EUR InStock
PRSI90-340
Human recombinant PARP1 (from Insect cells (Sf21))
Protéines et Peptides
PARP-1 (ARTD1) is involved in the base excision repair (BER) pathway, by catalyzing the poly(ADP-ribosyl)ation of a limited number of acceptor proteins involved in chromatin architecture and in DNA metabolism. This modification follows DNA damages and appears as an obligatory step in a detection/signalling pathway leading to the reparation of DNA strand breaks. PARP-1 positively regulates the transcription of MTUS1 and negatively regulates the transcription of MTUS2/TIP150. It forms a complex with EEF1A1 and TXK that acts as a T-helper 1 (Th1) cell-specific transcription factor and binds the promoter of IFN-gamma to directly regulate its transcription, and is thus involved importantly in Th1 cytokine production.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité