Mouse recombinant CREB-binding highly active (from insect cells (Sf21))
Fournisseur: ProSci Inc.
90-344
PRSI90-344EA 488 EUR InStock
PRSI90-344
Mouse recombinant CREB-binding highly active (from insect cells (Sf21))
Protéines et Peptides
CREB-binding protein acetylates histones, giving a specific tag for transcriptional activation. It also acetylates non-histone proteins, like NCOA3 and FOXO1. It binds specifically to phosphorylated CREB and enhances its transcriptional activity toward cAMP-responsive genes. Acts as a coactivator of ALX1 in the presence of EP300.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité