Human Recombinant GCN5 highly active (from Insect cells (Sf21))
Fournisseur: ProSci Inc.
90-345
PRSI90-345EA 488 EUR InStock
PRSI90-345
Human Recombinant GCN5 highly active (from Insect cells (Sf21))
Protéines et Peptides
GCN5 functions as a histone acetyltransferase (HAT) to promote transcriptional activation. Acetylation of histones gives a specific tag for epigenetic transcription activation. It has significant histone acetyltransferase activity with core histones, but not with nucleosome core particles. In case of HIV-1 infection, it is recruited by the viral protein Tat. It regulates Tat's transactivating activity and may help inducing chromatin remodelling of proviral genes. GCN5 is a component of the Ada Two-A containing (ATAC) complex, a complex with histone acetyltransferase activity on histones H3 and H4.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité