Human recombinant TIP60 (highly active) (from insect cells (Sf21))
Fournisseur: ProSci Inc.
90-347
PRSI90-347EA 488 EUR InStock
PRSI90-347
Human recombinant TIP60 (highly active) (from insect cells (Sf21))
Protéines et Peptides
TIP60 is the catalytic subunit of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of selected genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome-DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex is required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis and DNA repair. NuA4 may also play a direct role in DNA repair when recruited to sites of DNA damage. TIP60 directly acetylates and activates ataxia-telangiectasia mutated (ATM). In case of HIV-1 infection, interaction with the viral Tat protein leads to KAT5 polyubiquitination and targets it to degradation.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité