Human recombinant NAD kinase (highly active) (from E. coli)
Fournisseur: ProSci Inc.
90-353
PRSI90-353EA 584 EUR InStock
PRSI90-353
Human recombinant NAD kinase (highly active) (from E. coli)
Enzymes
NAD kinase catalyses the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which in its reduced form acts as an electron donor for biosynthetic reactions. NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité