Human recombinant Kynurenine Aminotransferase II (from E. coli)
Fournisseur: ProSci Inc.
90-360
PRSI90-360EA 690 EUR InStock
PRSI90-360
Human recombinant Kynurenine Aminotransferase II (from E. coli)
Enzymes
Kynurenine aminotransferases (KATs) are pyridoxal-5’-phosphate-dependent enzymes that catalyse the conversion of L-kynurenine into kynurenic acid, a neuroactive metabolite whose unbalancing is associated with a number of brain disorders. Biochemical and structural investigations revealed that L-kynurenine (L-KYN) recognition by hKAT II is achieved by exploiting structural features that are peculiar of this isoform, thus offering the possibility to select/design inhibitor molecules specifically targeting hKAT II to be used as modulators of kynurenic acid synthesis in the CNS. hKAT II is one of the aminotransferases involved in the pyridoxal 5'-phosphate (PLP)-dependent irreversible transamination of L-kynurenine (L-KYN) to kynurenic acid (KYNA) in the central nervous system. When assayed in vitro the protein also displays beta-elimination activity.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité