Recombinant Zmp1 M. tuberculosis (from E. coli)
Fournisseur: ProSci Inc.
90-361
PRSI90-361EA 690 EUR InStock
PRSI90-361
Recombinant Zmp1 M. tuberculosis (from E. coli)
Protéines et Peptides
M13 endopeptidases are single-pass, transmembrane zinc-metallopeptidases responsible in human for the regulation of the biological activity of many hormones and peptides and are involved in many important processes such as blood pressure regulation (Neprilysin or NEP1), cardiovascular development (Endothelin Converting Enzyme-1 or ECE-12), prevention of haemolytic reaction (KELL) and phosphate homeostasis (PHEX). Zmp1 is a soluble enzyme member of the M13 endopeptidases family that plays a key role in Mycobacterium tuberculosis pathogenicity with a still unknown mechanism. Recently, it was shown to suppress inflammasome activation by inhibiting caspase-1 activation, thus preventing processing of pro-IL-1beta into IL-1beta and the consequent phagosome maturation.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité