Recombinant NAD Kinase B. subtilis (from E. coli)
Fournisseur: ProSci Inc.
90-364
PRSI90-364EA 654 EUR InStock
PRSI90-364
Recombinant NAD Kinase B. subtilis (from E. coli)
Enzymes
NAD kinase catalyses the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which in its reduced form acts as an electron donor for biosynthetic reactions. NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis. NAD kinase from Bacillus subtilis is not ATP-dependent but has a broader substrate specificity than human NAD kinase. It can economically produce NADP+ by using other nucleoside triphosphates as well as inorganic polyphosphate as a source of phosphorus. Catalytic activity: ATP + NAD+ = ADP + NADP+.
Fusion-Tag: Tag Free
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité