B. subtilis recombinant NAD synthetase (from E. coli)
Fournisseur: ProSci Inc.
90-367
PRSI90-367EA 690 EUR InStock
PRSI90-367
B. subtilis recombinant NAD synthetase (from E. coli)
Enzymes
NAD synthetase is an essential enzyme involved in both the de novo biosynthesis and salvage of NAD+, catalyzing the final step of both pathways. Since NAD has a vital role in cell metabolism, the enzyme represents a valid target for the development of new antimycobacterial agents. It was shown to be induced by heat shock, salt stress, oxidative stress, glucose limitation and oxygen limitation and is involved in biological processes like stress response and sporulation resulting in formation of a cellular spore. Catalytic activity: ATP + deamido-NAD+ + NH3 = AMP + diphosphate + NAD+
Fusion-Tag: Tag Free
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité