Mouse recombinant B7-H4 (from CHO cells)
Fournisseur: ProSci Inc.
90-371
PRSI90-371EA 584 EUR InStock
PRSI90-371
Mouse recombinant B7-H4 (from CHO cells)
Protéines et Peptides
B7-H4 is a B7 family member that negatively regulates T cell immunity by inhibiting T cell proliferation, cytokine production and cell cycle progression. In vitro, B7-H4 inhibits CD4+ and CD8+ T cell proliferation, cytokine production and generation of alloreactive cytotoxic T cells (CTLs). In vivo, blockade of endogenous B7-H4 by specific monoclonal antibody promotes T cell responses. B7-H4 ia an important negative regulator of innate immunity through growth inhibition of neutrophils. B7-H4 is expressed on some tumour cancer cells. The role of B7-H4 in tumour progression may be to transform precancerous cells and then protect them from immunosurveillance.
  • This recombinant proteins is for research use only.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité