Human Recombinant CCL5 (from E. coli)
Fournisseur: ProSci Inc.
90-383
PRSI90-383EA 884 EUR InStock
PRSI90-383
Human Recombinant CCL5 (from E. coli)
Protéines et Peptides
CCL5, also known as RANTES (Regulated upon Activation Normal T cell Expressed and Secreted), is an 8kDa β-chemokine that plays a primary role in the inflammatory immune response by means of its ability to attract and activate leukocytes. Human and mouse RANTES exhibit cross-species activity on human and mouse cells. CCL5 is secreted by many cell types at inflammatory sites and it exerts a wide range of activities through the receptors CCR1, CCR3, CCR4 and CCR5. In humans, CCR5 binding to CCL5 inhibits the infectivity of R5 (M-tropic) but not X4 (T-tropic) strains of HIV-1. The deposition of CCL5 on activated vascular endothelial cells is crucial for monocyte adhesion to damaged vasculature, but CCL5 oligomerisation is not required for the extravasation of adherent leukocytes. CCL5 is upregulated in breast cancer and promotes tumour progression through the attraction of proinflammatory macrophages in addition to its actions on tumour cells, stromal cells and the vasculature.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité