Human Recombinant CCL22 (from CHO cells)
Fournisseur: ProSci Inc.
90-385
PRSI90-385EA 509 EUR InStock
PRSI90-385
Human Recombinant CCL22 (from CHO cells)
Protéines et Peptides
CCL22, also named stimulated T cell chemotactic protein (STCP1), is a CC chemokine initially isolated from clones of monocyte-derived macrophages and binds to the receptor CCR4. Human CCL22 is highly expressed in macrophage and in monocyte-derived dendritic cells and thymus. It is also found in lymph node, appendix, activated monocytes, resting and activated macrophages. May play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. CCL22 has been shown to induce chemotaxis or Ca2+ mobilisation in dendritic cells, IL-2 activated NK cells and activated T lymphocytes.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité