Mouse recombinant CCL22 (from CHO cells)
Fournisseur: ProSci Inc.
90-387
PRSI90-387EA 528 EUR InStock
PRSI90-387
Mouse recombinant CCL22 (from CHO cells)
Protéines et Peptides
CCL22 (C-C Motif Chemokine 22; Stimulated T cell chemotactic protein; STCP1) is a CC chemokine initially isolated from clones of monocyte-derived macrophages and binds to the receptor CCR4. Human CCL22 is highly expressed in macrophage and in monocyte-derived dendritic cells and thymus. It is also found in lymph node, appendix, activated monocytes, resting and activated macrophages. May play a role in the trafficking of activated/effector T lymphocytes to inflammatory sites and other aspects of activated T lymphocyte physiology. CCL22 has been shown to induce chemotaxis or Ca2+ mobilisation in dendritic cells, IL-2 activated NK cells and activated T lymphocytes.
  • This recombinant proteins is for research use only.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité