Mouse recombinant CD54 non-lytic (from CHO cells)
Fournisseur: ProSci Inc.
90-397
PRSI90-397EA 448 EUR InStock
PRSI90-397
Mouse recombinant CD54 non-lytic (from CHO cells)
Protéines et Peptides
Intercellular Adhesion Molecule-1 (ICAM-1, CD54) binds the leukocyte integrins LFA-1 and Mac1. CD54 expression is weak on leukocytes, epithelial and resting endothelial cells, as well as some other cell types, but expression can be stimulated by IFN-gamma, TNF-alpha, IL-1beta and LPS. Soluble CD54 is found in a biologically active form in serum, probably as a result of proteolytic cleavage from the cell surface, and is elevated in patients with various inflammatory syndromes such as septic shock, LAD, cancer and transplantation.
  • This recombinant proteins is for research use only.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité