Human Recombinant CD137 (from E. coli)
Fournisseur: ProSci Inc.
90-410
PRSI90-410EA 588 EUR InStock
PRSI90-410
Human Recombinant CD137 (from E. coli)
Protéines et Peptides
Human CD137 (4-1BB) is a costimulatory molecule of the tumour necrosis factor (TNF) receptor superfamily. The glycoprotein 4-1BB is expressed mainly on activated CD4+ and CD8+ T cells and binds to a high-affinity ligand (4-1BBL) expressed on several antigen-presenting cells such as macrophages and activated B cells. Upon ligand binding, 4-1BB is associated with the tumour receptor-associated factors (TRAF), the adaptor protein and mediates downstream signalling events including the activation of NF-kappaB and cytokine production. 4-1BB signalling either by binding to 4-1BBL or by antibody ligation delivers signals for T cell activation and growth as well as monocyte proliferation and B cell survival, and plays a important role in the amplification of T cell-mediated immune responses. In contrast, it can also enhance activation-induced T cell apoptosis when triggered by engagement of the TCR/CD3 complex. In addition, the 4-1BB/4-1BBL costimulatory pathway has been shown to augment secondary CTL responses to several viruses and increase antitumour immunity. 4-1BB is therefore a promising candidate for immunotherapy of human cancer.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité