Human Recombinant CD160 (from CHO cells)
Fournisseur: ProSci Inc.
90-420
PRSI90-420EA 654 EUR InStock
PRSI90-420
Human Recombinant CD160 (from CHO cells)
Protéines et Peptides
CD160 is a GPI-anchored lymphocyte surface receptor in which expression is mostly restricted to the highly cytotoxic CD56(dim) CD16(+) peripheral blood NK subset. CD160 is a receptor showing broad specificity for both classical and non-classical MHC class I molecules. CD160 acts as a co-activator receptor for CD3-induced proliferation of CD4+ CD160+ T cells isolated from inflammatory skin lesions. CD160-Fc fusion protein targets a novel costimulatory pathway and prolongs allograft survival.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité