Human recombinant CD275 (from CHO cells)
Fournisseur: ProSci Inc.
90-433
PRSI90-433EA 654 EUR InStock
PRSI90-433
Human recombinant CD275 (from CHO cells)
Protéines et Peptides
Human B7-H2 is a member of the growing B7 family of immune costimulatory proteins. Other family members include B7-1, B7-2, B7-H1 (PD-L1), PD-L2 and B7-H3. B7 proteins are members of the immunoglobulin (Ig) superfamily, the extracellular domains contain 2 Ig-like domains and all members have short cytoplasmic domains. B7-H2 has been identified as the ligand for ICOS, a member of the CD28 family of costimulatory receptors. Human B7-H2 is expressed constitutively on resting B cells, dendritic cells and at low levels on monocytes. The B7-H2/ICOS interaction appears to play roles in T cell dependent B cell activation and Th differentiation.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité