Mouse recombinant CD275 (from CHO cells)
Fournisseur: ProSci Inc.
90-436
PRSI90-436EA 654 EUR InStock
PRSI90-436
Mouse recombinant CD275 (from CHO cells)
Protéines et Peptides
Mouse B7-H2, also called CD275, B7RP-1, B7h, LICOS and GL50, is a member of the growing B7 family of immune costimulatory proteins. CD275 has been identified as the ligand for ICOS, a member of the CD28 family of co-stimulatory receptors. Mouse CD275 is expressed on resting B cells and at low levels on monocytes. The CD275/ ICOS interaction appears to play roles in T cell dependent B cell activation and Th differentiation.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité