Human recombinant CD278 (from CHO cells)
Fournisseur: ProSci Inc.
90-445
PRSI90-445EA 584 EUR InStock
PRSI90-445
Human recombinant CD278 (from CHO cells)
Protéines et Peptides
CD278, also called inducible costimulator (ICOS) or CRP1 (CD28-related protein-1), is a member of the growing CD28 family of immune costimulatory receptors. Other family members are CD28, CTLA-4 and PD-1. ICOS is expressed on most CD45RO + cells. ICOS expression is upregulated within approximately 24-48 hours of activation on Th primed cells. B7-H2, a member of the B7 family of costimulatory ligands, has been identified as the ICOS ligand. The B7-H2/ICOS interaction appears to play roles in T cell dependent B cell activation and Th differentiation.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité