Human recombinant DcR3 (from CHO cells)
Fournisseur: ProSci Inc.
90-454
PRSI90-454EA 686 EUR InStock
PRSI90-454
Human recombinant DcR3 (from CHO cells)
Protéines et Peptides
Decoy receptor 3 (DcR3; TNFRSF6B; M68) is a member of the TNF receptor superfamily. DcR3 shares sequence identity with OPG (31%), TNF-RII (29%) and Fas (17%) and is expressed in a variety of different tissues and at high levels in many malignant tumours. Ligands of DcR3 include Fas ligand, homologous to lymphotoxin showing inducible expression and competing with HSV glycoprotein D for herpes virus entry mediator (a receptor expressed by T lymphocytes), TNF-like molecule 1A and heparan sulfate proteoglycans. DcR3 modulates the function of T cells, dendritic cells and macrophages. DcR3/Fc fusion proteins can bind to human and mouse B cells and suppress the activation of B cells.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité