Rat Recombinant HMGB1 (from CHO cells)
Fournisseur: ProSci Inc.
90-458
PRSI90-458EA 734 EUR InStock
PRSI90-458
Rat Recombinant HMGB1 (from CHO cells)
Protéines et Peptides
HMGB1 was originally discovered as an essential DNA-binding protein for regulating p53, NF-kappaB and other important proteins. It is secreted from activated dentric cells, macrophage and nectrotic cells, and acts as a ligand for RAGE, TLR-2 and TLR-4 expressed on surrounding cells. As a result, HMGB1 activates Rac, CDC42 and NF-kappaB inducing localised innate immunity of damaged tissue, tissue regeneration by recruitment of stem cells and hemostasis by induction of tissue factor expression. HMGB1 is also causative agent of various diseases as it causes localised inflammation such as arteriosclerosis, chronic rheumatoid arthritis and nephritis.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité