Human recombinant IL1B (from E. coli)
Fournisseur: ProSci Inc.
90-463
PRSI90-463EA 950 EUR InStock
PRSI90-463
Human recombinant IL1B (from E. coli)
Protéines et Peptides
IL-1beta is produced by activated macrophages. IL-1beta stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity. IL-1beta belongs to the IL-1 family of proteins that are involved in the inflammatory response. IL-1beta in most situations requires the inflammasome complex to be cleaved and secreted.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité