Mouse recombinant IL-4 non-lytic (from CHO cells)
Fournisseur: ProSci Inc.
90-478
PRSI90-478EA 448 EUR InStock
PRSI90-478
Mouse recombinant IL-4 non-lytic (from CHO cells)
Protéines et Peptides
Interleukin-4 (IL-4) is a cytokine produced by type 2 helper T cells, the Th2 cells. These cells tends to make a specific set of lymphokines including IL-4, IL-5, IL-6, IL-10, IL-13, IL-3 and GM-CSF and fail to produce IL-2, IFN-gamma, and lymphotoxin (TNF-beta). In addition, mast cells can produce IL-4. IL-4 exerts numerous effects on various hematopoietic cell types. On B cells, IL-4 promotes immunoglobulin class switching to IgE and IgG1 isotypes and upregulates MHC class II and CD23 expression. IL-4 promotes survival, growth, and differentiation of both T and B lymphocytes, mast cells and endothelial cells. In addition, IL-4 inhibits the production of TNF, IL-1, and IL-6 by macrophages.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité