Mouse recombinant IL-6 non-lytic (from CHO cells)
Fournisseur: ProSci Inc.
90-486
PRSI90-486EA 509 EUR InStock
PRSI90-486
Mouse recombinant IL-6 non-lytic (from CHO cells)
Protéines et Peptides
Interleukin 6 (IL-6) is a pleiotropic alpha-helical cytokine that plays important roles in acute phase reactions, inflammation, hematopoiesis, bone metabolism and cancer progression. IL-6 activity is essential for the transition from acute inflammation to either acquired immunity or chronic inflammatory disease. It is secreted by multiple cell types as a 22kDa-28kDa phosphorylated and variably glycosylated molecule. IL-6 induces signallling through a cell surface heterodimeric receptor complex composed of a ligand binding subunit (IL-6R) and a signal transducing subunit (gp130). IL-6 is a key factor for the growth of plasma cells.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité