Mouse recombinant IL6 Receptor (from CHO cells)
Fournisseur: ProSci Inc.
90-488
PRSI90-488EA 620 EUR InStock
PRSI90-488
Mouse recombinant IL6 Receptor (from CHO cells)
Protéines et Peptides
Interleukin 6 receptor (IL-6R; CD126) is a type I cytokine receptor, which is a protein complex consisting of an IL-6 receptor subunit (IL-6R) and interleukin 6 signal transducer glycoprotein 130 (gp130). It is a potent pleiotropic cytokine that regulates cell growth and differentiation and plays an important role in immune response. Dysregulated production of IL-6 and IL-6R are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité