Human recombinant IL-15 mutant (from CHO cells), Biotin
Fournisseur: ProSci Inc.
90-505
PRSI90-505EA 734 EUR InStock
PRSI90-505
Human recombinant IL-15 mutant (from CHO cells), Biotin
Protéines et Peptides
Interleukin-15 (IL-15) has a broad spectrum of biological activities. It is crucial for the development, proliferation, survival and differentiation of multiple cells from both innate and adaptive immune systems. IL-15 up-regulation has a central role in the development of several autoimmune or chronic inflammatory disorders. Targeting IL-15 or its receptor may have a valuable impact on the treatment of immune-mediated diseases. IL-15 participates in the development of important immune antitumour mechanisms. It activates CD8(+) T cells, natural killer (NK) cells, NK T cells, and can promote the formation of antitumour antibodies. IL-15 can also protect T effector cells from the action of T regulatory cells and reverse tolerance to tumour-associated antigens. In pre-clinical studies IL-15 has been found to demonstrate potentiated antitumour effects following pre-association with IL-15Ralpha, or when used in combination with chemotherapy, adoptive therapy, monoclonal antibodies, and tumour vaccines.
Application: Useful for immunofluorescent staining and flow cytometric analysis to identify and enumerate IL-15Ralpha expressing cells within mixed cell populations.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité