Human recombinant IL18 (from E. coli)
Fournisseur: ProSci Inc.
90-506
PRSI90-506EA 377 EUR InStock
PRSI90-506
Human recombinant IL18 (from E. coli)
Protéines et Peptides
Interleukin-18 (IL-18) is a costimulatory factor for production of interferon-gamma (IFN-gamma) in response to toxic shock and shares functional similarities with IL-12. IL-18 is synthesised as a precursor 24kDa molecule without a signal peptide and must be cleaved to produce an active molecule. IL-1 converting enzyme (ICE; Caspase-1) cleaves pro-IL-18 at aspartic acid in the P1 position, producing the mature, bioactive peptide that is readily released from the cells. It is reported that IL-18 is produced from Kupffer cells, activated macrophages, keratinocytes, intestinal epithelial cells, osteoblasts, adrenal cortex cells and murine diencephalon. IFN-gamma is produced by activated T or NK cells and plays critical roles in the defense against microbiral pathogens. IFN-gamma activates macrophages and enhances NK activity and B cell maturation, proliferation and Ig secretion. IFN-gamma also induces expression of MHC class I and II antigens and inhibits osteoclast activation. IL-18 acts on T helper type-1 (Th1) T cells and in combination with IL-12 strongly induces them to produce IFN-gamma. Pleiotropic effects of IL-18 have also been reported, such as enhancement production of IFN-gamma and GM-CSF in peripheral blood mononuclear cells, production of Th1 cytokines, IL-2, GM-CSF, IFN-gamma in T cells and enhancement of Fas ligand expression by Th1 cells.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité