Human recombinant IL23R (from CHO cells)
Fournisseur: ProSci Inc.
90-526
PRSI90-526EA 584 EUR InStock
PRSI90-526
Human recombinant IL23R (from CHO cells)
Protéines et Peptides
Interleukin-23 receptor (IL-23R), is a member of the type I cytokine receptor family, type 2 subfamily. It is expressed by monocytes, Th1, Th0, NK and dendritic cells. Isoform 1 is specifically expressed in NK cells. IL-23 functions in innate and adaptive immunity and may participate in acute response to infection in peripheral tissues. IL-23 may be responsible for autoimmune inflammatory diseases and be important for tumourigenesis. IL-23R associates with IL-12RB1 to form the interleukin-23 receptor. It binds IL-23 and mediates T cells, NK cells and possibly certain macrophage/myeloid cells stimulation probably through activation of the Jak-Stat signalling cascade. Genetic variations in IL-23R are associated with inflammatory bowel disease type 17 (IBD17). IBD17 is a chronic, relapsing inflammation of the gastrointestinal tract with a complex etiology. Genetic variations in IL-23R also can cause susceptibility to psoriasis type 7.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité