Human recombinant IL24 (from CHO cells)
Fournisseur: ProSci Inc.
90-527
PRSI90-527EA 612 EUR InStock
PRSI90-527
Human recombinant IL24 (from CHO cells)
Protéines et Peptides
IL-24 is a cytokine belonging to the IL-10 family of cytokines that signals through two heterodimeric receptors: IL-20R1/IL-20R2 and IL-22R1/IL-20R2. This interleukin is also known as melanoma differentiation-associated 7 (mda-7) due to its discovery as a tumour suppressing protein. IL-24 appears to control cell survival and proliferation by inducing rapid activation of particular transcription factors called STAT1 and STAT3. It is predominantly released by activated monocytes, macrophages and T helper 2 (Th2) cells and acts on non-haematopoietic tissues such as skin, lung and reproductive tissues. IL-24 performs important roles in wound healing, psoriasis and cancer. IL-24 has antiproliferative properties is inhibiting angiogenesis and may contribute to terminal cell differentiation.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité