Mouse recombinant TIM-1 (from CHO cells)
Fournisseur: ProSci Inc.
90-547
PRSI90-547EA 773 EUR InStock
PRSI90-547
Mouse recombinant TIM-1 (from CHO cells)
Protéines et Peptides
The TIM (T cell/transmembrane, immunoglobulin and mucin) family plays a critical role in regulating immune responses, including allergy, asthma, transplant tolerance, autoimmunity and the response to viral infections. The unique structure of TIM immunoglobulin variable region domains allows highly specific recognition of phosphatidylserine (PtdSer), exposed on the surface of apoptotic cells. Tim-1 (KIM-1; HAVcr-1) is a 100 kDa, type I transmembrane glycoprotein member of the TIM family of immunoglobulin superfamily molecules. There are two cytoplasmic alternate splice forms of Tim-1. One is a long (359 aa) kidney form termed Tim-1b, and one is a short (334 aa) liver form termed Tim-1a. Tim-1, important for asthma and allergy, is preferentially expressed on T-helper 2 (Th2) cells and functions as a potent costimulatory molecule for T cell activation.
Fusion-Tag: Fc Tag
This recombinant proteins is for research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité