Human recombinant IFN alpha 2b (from E. coli)
Fournisseur: ProSci Inc.
91-004
PRSI91-004EA 409 EUR InStock
PRSI91-004
Human recombinant IFN alpha 2b (from E. coli)
Protéines et Peptides
At least 23 different variants of IFN- alpha are known. The individual proteins have molecular masses between 19-26 kDa and consist of proteins with lengths of 156-166 and 172 amino acids. All IFN- alpha subtypes possess a common conserved sequence region between amino acid positions 115-151 while the amino-terminal ends are variable. Many IFN- alpha subtypes differ in their sequences by only one or two positions. Naturally occurring variants also include proteins that are truncated by 10 amino acids at the carboxyl-terminal end.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité