Human recombinant IL11 (from P. pichia)
Fournisseur: ProSci Inc.
91-005
PRSI91-005EA 852 EUR InStock
PRSI91-005
Human recombinant IL11 (from P. pichia)
Protéines et Peptides
Interleukin 11 (IL-11) is a thrombopoietic growth factor that directly stimulates the proliferation of hematopoietic stem cells and megakaryocyte progenitor cells and induces megakaryocyte maturation resulting in increased platelet production. IL-11 is a member of a family of human growth factors that includes human growth hormone, granulocyte colony-stimulating factor, and other growth factors.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité