Human recombinant TNF alpha (from E. coli)
Fournisseur: ProSci Inc.
91-006
PRSI91-006EA 336 EUR InStock
PRSI91-006
Human recombinant TNF alpha (from E. coli)
Protéines et Peptides
TNF alpha is a homotrimer with a subunit molecular mass of 17 kD and plays a major role in growth regulation, differentiation, inflammation, viral replication, tumorigenesis, autoimmune diseases and in viral, bacterial, fungal, and parasitic infections. Besides inducing hemorrhagic necrosis of tumors, TNF was found to be involved in tumorigenesis, tumor metastasis, viral replication, septic shock, fever, inflammation, and autoimmune diseases including Crohn’s disease, and rheumatoid arthritis as well as graft-versus-host disease.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité