Human recombinant TRAIL (from E. coli)
Fournisseur: ProSci Inc.
91-016
PRSI91-016EA 409 EUR InStock
PRSI91-016
Human recombinant TRAIL (from E. coli)
Protéines et Peptides
Human TNFSF10 is a type II transmembrane protein with an intracellular N-terminus and a ‘TNF homology domain’ (THD) at the extracellular C terminus. TNFSF10 can interact with several distinct receptors. Two of these receptors that belongs to TNFR superfamily, DR4 (TRAIL-R1) and DR5 (TRAIL-R2/TRICK2), are plasma membrane proteins containing intracellular death domains essential for activating apoptosis. TNFSF10 is promising for cancer therapy because it is cytotoxic and activates apoptosis in the majority of malignant cells, but not in normal cells.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité