Human Recombinant EGF (from E. coli)
Fournisseur: ProSci Inc.
91-020
PRSI91-020EA 343 EUR InStock
PRSI91-020
Human Recombinant EGF (from E. coli)
Protéines et Peptides
Epidermal growth factor (EGF) is a small mitogenic protein that is thought to be involved in mechanisms such as normal cell growth, oncogenesis, and wound healing. This protein shows both strong sequential and functional homology with human type-alpha transforming growth factor (hTGF alpha), which is a competitor for EGF receptor sites. EGF is a small 53 amino acid residue long protein that contains three disulfide bridges
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité