Human recombinant IGFI (from E. coli)
Fournisseur: ProSci Inc.
91-022
PRSI91-022EA 343 EUR InStock
PRSI91-022
Human recombinant IGFI (from E. coli)
Protéines et Peptides
Insulin-like growth factor I (IGF1) belongs to the family of insulin-like growth factors that are structurally homologous to proinsulin. Mature IGFs are generated by proteolytic processing of inactive precursor protein containing N-terminal and C-terminal propeptide regions. Mature human IGF-I consisting of 70 amino acids with 94% identity with mouse IGF1 and exhibits cross-species activity. IGF1 binds IGF-1R, IGF-2R, and the insulin receptor and plays a key role in cell cycle progression, cell proliferation and tumor progression. IGF1 expression is regulated by growth hormone.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité