Human recombinant GMCSF (from P. pichia)
Fournisseur: ProSci Inc.
91-030
PRSI91-030EA 654 EUR InStock
PRSI91-030
Human recombinant GMCSF (from P. pichia)
Protéines et Peptides
GM-CSF was initially characterized as a growth factor that can support the in vitro colony formation of granulocyte macrophage progenitors. It is produced by a number of different cell types (including activated T cells, B cells, macrophages, mast cells, endothelial cells and fibroblasts) in response to cytokine of immune and inflammatory stimuli. Besides granulocyte-macrophage progenitors, GM-CSF is also a growth factor for erythroid, megakaryocyte and eosinophil progenitors. On mature hematopoietic, monocytes/macrophages, and eosinophils, GM-CSF has also been reported to have a functional role on non-hematopoitic cells. It can induce human endothelial cells to migrate and proliferate. Additionally, GM-CSF can also stimulate the proliferation of a number of tumor cell lines, including osteogenic sarcoma, carcinoma and adenocarcinoma cell lines.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité